missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)phenol, 98%, Thermo Scientific™

$132.22 - $132.22

Chemical Identifiers

CAS 827-99-6
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040987
InChI Key UWLJERQTLRORJN-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
PubChem CID 2733261
IUPAC Name 3-(trifluoromethoxy)phenol
SMILES OC1=CC=CC(OC(F)(F)F)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC308700010
SDS View Documents
Thermo Scientific Chemicals
308700010
1 g Glass bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

827-99-6
178.11
UWLJERQTLRORJN-UHFFFAOYSA-N
2733261
OC1=CC=CC(OC(F)(F)F)=C1
C7H5F3O2
MFCD00040987
3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
3-(trifluoromethoxy)phenol

Specifications

827-99-6
100.0
1.3700g/mL
84°C
97.5% min. (GC)
C7H5F3O2
CF3OC6H4OH
1 g
3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
OC1=CC=CC(OC(F)(F)F)=C1
178.11
178.11
Liquid
97.5
Colorless to Yellow
69.0°C to 70.0°C (12.0 mmHg)
Authentic
Glass bottle
1.4450 to 1.4470
MFCD00040987
1.37
UWLJERQTLRORJN-UHFFFAOYSA-N
3-(trifluoromethoxy)phenol
2733261
98%
3-(Trifluoromethoxy)phenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only