missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzyl Bromide 97.0+%, TCI America™

Catalog No. T27301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27301G 1 g
T27305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27301G Supplier TCI America Supplier No. T27301G

Chemical Identifiers

CAS 159689-88-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061271
InChI Key QSIVWRRHVXSDNE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-trifluoromethoxy benzene, 3-trifluoromethoxybenzyl bromide, alpha-bromo-3-trifluoromethoxy toluene, 3-trifluoromethoxy benzylbromide, m-trifluoromethoxy benzyl bromide, 3-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-3-trifluoromethoxy-benzene, benzene, 1-bromomethyl-3-trifluoromethoxy
PubChem CID 737176
IUPAC Name 1-(bromomethyl)-3-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC(CBr)=C1

Specifications

Color Yellow
Boiling Point 179°C
Quantity 1 g
UN Number 3265
Formula Weight 255.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)benzyl Bromide