missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzyl bromide, 98%

Catalog No. AAB2506106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2506106 5 g
AAB2506103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2506106 Supplier Thermo Scientific Chemicals Supplier No. B2506106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 159689-88-0 | C8H6BrF3O | 255.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 159689-88-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061271
InChI Key QSIVWRRHVXSDNE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-trifluoromethoxy benzene, 3-trifluoromethoxybenzyl bromide, alpha-bromo-3-trifluoromethoxy toluene, 3-trifluoromethoxy benzylbromide, m-trifluoromethoxy benzyl bromide, 3-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-3-trifluoromethoxy-benzene, benzene, 1-bromomethyl-3-trifluoromethoxy
PubChem CID 737176
IUPAC Name 1-(bromomethyl)-3-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC(CBr)=C1

Specifications

Density 1.572
Boiling Point 178°C to 180°C
Flash Point 88°C (190°F)
Refractive Index 1.478
Quantity 5 g
UN Number UN3265
Beilstein 8052203
Formula Weight 255.04
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)benzyl bromide
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only