missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzonitrile 98.0+%, TCI America™

Catalog No. T23721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23721G 1 g
T23725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23721G Supplier TCI America Supplier No. T23721G
Only null left

Chemical Identifiers

CAS 52771-22-9
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00040959
InChI Key DCZAPXGEZYVQNX-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzonitrile, m-trifluoromethoxybenzonitrile, benzonitrile, 3-trifluoromethoxy, 3-trifluoromethoxy-benzonitrile, 3-trifluoromethoxy benzenecarbonitrile, pubchem4825, acmc-1art8, ksc493e1j, m-trifluoromethoxy benzonitrile
PubChem CID 142962
IUPAC Name 3-(trifluoromethoxy)benzonitrile
SMILES FC(F)(F)OC1=CC=CC(=C1)C#N

Specifications

Color Yellow
Quantity 1 g
UN Number 3276
Formula Weight 187.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)benzonitrile