missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7137189
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23831G 1 g
T23835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23831G Supplier TCI America Supplier No. T23831G
Only null left

Chemical Identifiers

CAS 1014-81-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00041501
InChI Key OKPFIWIMBJNFSE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzoic acid, m-trifluoromethoxybenzoic acid, m-trifluoromethoxy benzoic acid, benzoic acid, 3-trifluoromethoxy, 3-trifluoromethoxy benzoicacid, alpha,alpha,alpha-trifluoro-m-anisic acid, 3-carboxy-alpha,alpha,alpha-trifluoroanisole, 3-trifluoromethoxy-benzoic acid, pubchem4590, acmc-1btyb
PubChem CID 66096
IUPAC Name 3-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 1 g
Formula Weight 206.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethoxy)benzoic Acid