missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. T35085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35085G 5 g
T35081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35085G Supplier TCI America Supplier No. T35085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 220227-84-9
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.611
MDL Number MFCD01091016
InChI Key DODDSXTWDSJCDN-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzenesulfonyl chloride, 3-trifluoromethoxy benzene-1-sulfonyl chloride, 3-trifluoromethoxy benzenesulphonyl chloride, 3-trifluoromethoxy-benzenesulfonyl chloride, 3-trifluoromethoxybenzenesulfonylchloride, 3-trifluoromethyl oxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3-trifluoromethoxy, pubchem5187, acmc-1ce4e
PubChem CID 2777218
IUPAC Name 3-(trifluoromethoxy)benzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 230°C
Quantity 5 g
UN Number 3265
Formula Weight 260.61
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)benzenesulfonyl Chloride