missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)aniline 98.0+%, TCI America™

Catalog No. T22551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22551G 1 g
T22555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22551G Supplier TCI America Supplier No. T22551G

Chemical Identifiers

CAS 1535-73-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
InChI Key SADHVOSOZBAAGL-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
PubChem CID 73753
IUPAC Name 3-(trifluoromethoxy)aniline
SMILES C1=CC(=CC(=C1)OC(F)(F)F)N

Specifications

Boiling Point 70°C
Quantity 1 g
Formula Weight 177.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)aniline