missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)aniline, 98%

CAS: 1535-73-5 | C7H6F3NO | 177.126 g/mol

Supplier:  Thermo Scientific Chemicals A1959206

 View more versions of this product

Catalog No. AAA1959206


Only null left
Explore available promotions

Description

Description

3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-(Trifluoromethoxy)aniline
1.325
85°C (185°F)
C7H6F3NO
MFCD00041511
UN2810
3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
SADHVOSOZBAAGL-UHFFFAOYSA-N
3-(trifluoromethoxy)aniline
73753
98%
1535-73-5
72°C to 75°C (8 mmHg)
Characteristic
1.466
5 g
776921
Not miscible or difficult to mix in water.
C1=CC(=CC(=C1)OC(F)(F)F)N
177.126
177.13

Chemical Identifiers

1535-73-5
177.126
SADHVOSOZBAAGL-UHFFFAOYSA-N
73753
C1=CC(=CC(=C1)OC(F)(F)F)N
C7H6F3NO
MFCD00041511
3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
3-(trifluoromethoxy)aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 216-256-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only