missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)aniline, 98%

Catalog No. AAA1959203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1959203 1 g
AAA1959206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1959203 Supplier Thermo Scientific Chemicals Supplier No. A1959203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1535-73-5 | C7H6F3NO | 177.126 g/mol

3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1535-73-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00041511
InChI Key SADHVOSOZBAAGL-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
PubChem CID 73753
IUPAC Name 3-(trifluoromethoxy)aniline
SMILES C1=CC(=CC(=C1)OC(F)(F)F)N

Specifications

Density 1.325
Boiling Point 72°C to 75°C (8 mmHg)
Flash Point 85°C (185°F)
Odor Characteristic
Refractive Index 1.466
Quantity 1 g
UN Number UN2810
Beilstein 776921
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 177.13
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)aniline
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 216-256-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only