missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethoxy)acetophenone 97.0+%, TCI America™

Catalog No. T15875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15875G 5 g
T15871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15875G Supplier TCI America Supplier No. T15875G

Chemical Identifiers

CAS 170141-63-6
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042403
InChI Key UYHTUQHYGKAYJM-UHFFFAOYSA-N
Synonym 3'-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethanone, 3-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethan-1-one, 3-trifluoromethoxyacetophenone, 1-acetyl-3-trifluoromethoxy benzene, fxffor cv1, pubchem4297, intermediates-zcf02268
PubChem CID 737161
IUPAC Name 1-[3-(trifluoromethoxy)phenyl]ethan-1-one
SMILES CC(=O)C1=CC=CC(OC(F)(F)F)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-(Trifluoromethoxy)acetophenone