CAS RN 170141-63-6
IUPAC Name:
1-[3-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms:
1-[3-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol):
204.15
Molecular Formula:
C9H7F3O2
InChi Key:
UYHTUQHYGKAYJM-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
3'-(Trifluoromethoxy)acetophenone 97.0+%, TCI America™
CAS: 170141-63-6 Molecular Formula: C9H7F3O2 Molecular Weight (g/mol): 204.15 MDL Number: MFCD00042403 InChI Key: UYHTUQHYGKAYJM-UHFFFAOYSA-N Synonym: 3'-trifluoromethoxy acetophenone,1-3-trifluoromethoxy phenyl ethanone,3-trifluoromethoxy acetophenone,1-3-trifluoromethoxy phenyl ethan-1-one,3-trifluoromethoxyacetophenone,1-acetyl-3-trifluoromethoxy benzene,fxffor cv1,pubchem4297,intermediates-zcf02268 PubChem CID: 737161 IUPAC Name: 1-[3-(trifluoromethoxy)phenyl]ethan-1-one SMILES: CC(=O)C1=CC=CC(OC(F)(F)F)=C1