missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoroacetyl)indole 98.0+%, TCI America™

Supplier:  TCI America T18091G

 View more versions of this product

Catalog No. T18091G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-(Trifluoroacetyl)indole
212°C
MFCD00182114
3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one
FC(F)(F)C(=O)C1=CNC2=CC=CC=C12
213.16
213.16
Crystalline Powder
14618-45-2
C10H6F3NO
1 g
LCMDCXWSHDFQKP-UHFFFAOYSA-N
2,2,2-trifluoro-1-(1H-indol-3-yl)ethan-1-one
589126
≥98.0% (GC)

Chemical Identifiers

14618-45-2
213.16
LCMDCXWSHDFQKP-UHFFFAOYSA-N
589126
FC(F)(F)C(=O)C1=CNC2=CC=CC=C12
C10H6F3NO
MFCD00182114
3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one
2,2,2-trifluoro-1-(1H-indol-3-yl)ethan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions