missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoroacetyl)indole 98.0+%, TCI America™
$100.67 - $287.06
Chemical Identifiers
| CAS | 14618-45-2 |
|---|---|
| Molecular Formula | C10H6F3NO |
| Molecular Weight (g/mol) | 213.16 |
| MDL Number | MFCD00182114 |
| InChI Key | LCMDCXWSHDFQKP-UHFFFAOYSA-N |
| Synonym | 3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one |
| PubChem CID | 589126 |
| IUPAC Name | 2,2,2-trifluoro-1-(1H-indol-3-yl)ethan-1-one |
| SMILES | FC(F)(F)C(=O)C1=CNC2=CC=CC=C12 |
Chemical Identifiers
| 14618-45-2 | |
| 213.16 | |
| LCMDCXWSHDFQKP-UHFFFAOYSA-N | |
| 589126 | |
| FC(F)(F)C(=O)C1=CNC2=CC=CC=C12 |
| C10H6F3NO | |
| MFCD00182114 | |
| 3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one | |
| 2,2,2-trifluoro-1-(1H-indol-3-yl)ethan-1-one |
Specifications
| 14618-45-2 | |
| C10H6F3NO | |
| 1 g | |
| LCMDCXWSHDFQKP-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-(1H-indol-3-yl)ethan-1-one | |
| 589126 | |
| ≥98.0% (GC) | |
| 3-(Trifluoroacetyl)indole |
| 212°C | |
| MFCD00182114 | |
| 3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one | |
| FC(F)(F)C(=O)C1=CNC2=CC=CC=C12 | |
| 213.16 | |
| 213.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No