missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenemethanol, 97%, Thermo Scientific™

$180.63 - $654.55

Chemical Identifiers

CAS 71637-34-8
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014534
InChI Key BOWIFWCBNWWZOG-UHFFFAOYSA-N
Synonym 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
PubChem CID 123570
IUPAC Name thiophen-3-ylmethanol
SMILES OCC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC442240050
View Documents Promotion Details
N/A 5g
Each for $180.63
Only null left
 
AC442240250
View Documents Promotion Details
N/A 25g
Each for $654.55
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

71637-34-8
114.16
BOWIFWCBNWWZOG-UHFFFAOYSA-N
123570
OCC1=CSC=C1
C5H6OS
MFCD00014534
3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
thiophen-3-ylmethanol

Specifications

71637-34-8
73.0°C (6.0 Torr)
96% min. (GC)
MFCD00014534
3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
OCC1=CSC=C1
114.16
114.17
3-Thiophenemethanol
1.2110g/mL
100.5°C
C5H6OS
1.211
BOWIFWCBNWWZOG-UHFFFAOYSA-N
thiophen-3-ylmethanol
123570
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 275-741-8

SDS
Documents

Documents

RUO – Research Use Only