missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenemethanol, 97%

Catalog No. AC442240050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442240050 5 g
AC442240250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442240050 Supplier Thermo Scientific Chemicals Supplier No. 442240050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 71637-34-8 | C5H6OS | 114.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71637-34-8
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014534
InChI Key BOWIFWCBNWWZOG-UHFFFAOYSA-N
Synonym 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
PubChem CID 123570
IUPAC Name thiophen-3-ylmethanol
SMILES OCC1=CSC=C1

Specifications

Density 1.2110g/mL
Boiling Point 73.0°C (6.0 Torr)
Flash Point 100.5°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.211
Formula Weight 114.17
Percent Purity 97%
Chemical Name or Material 3-Thiophenemethanol
EINECSNumber 275-741-8

RUO – Research Use Only