missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiophenemalonic Acid 98.0+%, TCI America™
Supplier: TCI America T135425G
Specifications
| 3-Thiophenemalonic Acid | |
| Yellow | |
| MFCD00005469 | |
| 3-thiophenemalonic acid, 3-thienylmalonic acid, 2-thiophen-3-yl malonic acid, propanedioic acid, 3-thienyl, 3-thiophene malonic acid, 3-thienylpropanedioic acid, 3-thienyl malonic acid, thiophene-3-malonic acid, 2-3-thienyl malonic acid, 2-3-thienyl propanedioic acid | |
| [O-]C(=O)C(C([O-])=O)C1=CSC=C1 | |
| 184.17 | |
| 186.18 | |
| Crystalline Powder |
| 21080-92-2 | |
| C7H4O4S | |
| 25 g | |
| GCOOGCQWQFRJEK-UHFFFAOYSA-L | |
| 2-(thiophen-3-yl)propanedioate | |
| 88782 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 21080-92-2 | |
| 184.17 | |
| GCOOGCQWQFRJEK-UHFFFAOYSA-L | |
| 88782 | |
| [O-]C(=O)C(C([O-])=O)C1=CSC=C1 |
| C7H4O4S | |
| MFCD00005469 | |
| 3-thiophenemalonic acid, 3-thienylmalonic acid, 2-thiophen-3-yl malonic acid, propanedioic acid, 3-thienyl, 3-thiophene malonic acid, 3-thienylpropanedioic acid, 3-thienyl malonic acid, thiophene-3-malonic acid, 2-3-thienyl malonic acid, 2-3-thienyl propanedioic acid | |
| 2-(thiophen-3-yl)propanedioate |
Safety and Handling
TSCA : Yes