missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiophenemalonic Acid 98.0+%, TCI America™
$41.02 - $128.62
Chemical Identifiers
| CAS | 21080-92-2 |
|---|---|
| Molecular Formula | C7H4O4S |
| Molecular Weight (g/mol) | 184.17 |
| MDL Number | MFCD00005469 |
| InChI Key | GCOOGCQWQFRJEK-UHFFFAOYSA-L |
| Synonym | 3-thiophenemalonic acid, 3-thienylmalonic acid, 2-thiophen-3-yl malonic acid, propanedioic acid, 3-thienyl, 3-thiophene malonic acid, 3-thienylpropanedioic acid, 3-thienyl malonic acid, thiophene-3-malonic acid, 2-3-thienyl malonic acid, 2-3-thienyl propanedioic acid |
| PubChem CID | 88782 |
| IUPAC Name | 2-(thiophen-3-yl)propanedioate |
| SMILES | [O-]C(=O)C(C([O-])=O)C1=CSC=C1 |
Chemical Identifiers
| 21080-92-2 | |
| 184.17 | |
| GCOOGCQWQFRJEK-UHFFFAOYSA-L | |
| 88782 | |
| [O-]C(=O)C(C([O-])=O)C1=CSC=C1 |
| C7H4O4S | |
| MFCD00005469 | |
| 3-thiophenemalonic acid, 3-thienylmalonic acid, 2-thiophen-3-yl malonic acid, propanedioic acid, 3-thienyl, 3-thiophene malonic acid, 3-thienylpropanedioic acid, 3-thienyl malonic acid, thiophene-3-malonic acid, 2-3-thienyl malonic acid, 2-3-thienyl propanedioic acid | |
| 2-(thiophen-3-yl)propanedioate |
Specifications
| 21080-92-2 | |
| C7H4O4S | |
| 5 g | |
| GCOOGCQWQFRJEK-UHFFFAOYSA-L | |
| 2-(thiophen-3-yl)propanedioate | |
| 88782 | |
| ≥98.0% (HPLC,T) | |
| 3-Thiophenemalonic Acid |
| Yellow | |
| MFCD00005469 | |
| 3-thiophenemalonic acid, 3-thienylmalonic acid, 2-thiophen-3-yl malonic acid, propanedioic acid, 3-thienyl, 3-thiophene malonic acid, 3-thienylpropanedioic acid, 3-thienyl malonic acid, thiophene-3-malonic acid, 2-3-thienyl malonic acid, 2-3-thienyl propanedioic acid | |
| [O-]C(=O)C(C([O-])=O)C1=CSC=C1 | |
| 184.17 | |
| 186.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes