Learn More
3-Thiopheneethanol, 98%, Thermo Scientific Chemicals
$176.18 - $176.18
Chemical Identifiers
| CAS | 13781-67-4 |
|---|---|
| Molecular Formula | C6H8OS |
| Molecular Weight (g/mol) | 128.19 |
| MDL Number | MFCD00009766 |
| InChI Key | YYPNNBPPDFTQFX-UHFFFAOYSA-N |
| Synonym | 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 |
| PubChem CID | 83731 |
| IUPAC Name | 2-thiophen-3-ylethanol |
| SMILES | OCCC1=CSC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1513806
|
Thermo Scientific Chemicals
L1513806 |
5 g |
Each for $176.18
|
|
|||||
Description
3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene
Solubility
Sparingly soluble in water(0.059 g/L at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Chemical Identifiers
| 13781-67-4 | |
| 128.19 | |
| YYPNNBPPDFTQFX-UHFFFAOYSA-N | |
| 83731 | |
| OCCC1=CSC=C1 |
| C6H8OS | |
| MFCD00009766 | |
| 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 | |
| 2-thiophen-3-ylethanol |
Specifications
| 13781-67-4 | |
| 110°C to 111°C (14 mmHg) | |
| C6H8OS | |
| MFCD00009766 | |
| 107080 | |
| Sparingly soluble in water(0.059g/L at 25°C). | |
| OCCC1=CSC=C1 | |
| 128.19 | |
| 128.19 | |
| 3-Thiopheneethanol |
| 1.159 | |
| 92°C (197°F) | |
| 1.553 | |
| 5 g | |
| 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 | |
| YYPNNBPPDFTQFX-UHFFFAOYSA-N | |
| 2-thiophen-3-ylethanol | |
| 83731 | |
| 98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P233-P240-P241-P242-P243-P280-P370+P378q-P374-P501c
H227
EINECSNumber : 237-434-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only