missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7136599
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T10845G 5 g
T108425G 25 g
2 options

Catalog No. T10845G

Supplier: TCI America T10845G

Only null left

Chemical Identifiers

CAS 88-13-1
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.145
MDL Number MFCD00005467
InChI Key YNVOMSDITJMNET-UHFFFAOYSA-N
Synonym 3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be
PubChem CID 6918
IUPAC Name thiophene-3-carboxylic acid
SMILES C1=CSC=C1C(=O)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 128.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Thiophenecarboxylic Acid
RTECSNumber XM8330300
TSCA No