missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiophenecarboxaldehyde 98.0+%, TCI America™
Supplier: TCI America T086425G
Specifications
| 3-Thiophenecarboxaldehyde | |
| Yellow | |
| C5H4OS | |
| 25 g | |
| RBIGKSZIQCTIJF-UHFFFAOYSA-N | |
| thiophene-3-carbaldehyde | |
| 68135 | |
| 112.15 | |
| Liquid |
| 498-62-4 | |
| 198°C | |
| MFCD00005466 | |
| 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde | |
| C1=CSC=C1C=O | |
| 112.146 | |
| CHEBI:87611 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 498-62-4 | |
| 112.146 | |
| RBIGKSZIQCTIJF-UHFFFAOYSA-N | |
| 68135 | |
| thiophene-3-carbaldehyde |
| C5H4OS | |
| MFCD00005466 | |
| 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde | |
| CHEBI:87611 | |
| C1=CSC=C1C=O |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator