missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarboxaldehyde 98.0+%, TCI America™

Catalog No. p-7136465
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T08645G 5 g
T086425G 25 g
2 options

Catalog No. T08645G

Supplier: TCI America T08645G

Only null left

Chemical Identifiers

CAS 498-62-4
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.146
MDL Number MFCD00005466
InChI Key RBIGKSZIQCTIJF-UHFFFAOYSA-N
Synonym 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
PubChem CID 68135
ChEBI CHEBI:87611
IUPAC Name thiophene-3-carbaldehyde
SMILES C1=CSC=C1C=O

Specifications

Color Yellow
Boiling Point 198°C
Quantity 5 g
Formula Weight 112.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Thiophenecarboxaldehyde
TSCA Yes
Recommended Storage Refrigerator