missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiopheneacetic acid, 98%
Description
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
Specifications
| Melting Point | 77°C to 81°C |
| Boiling Point | 129°C (2.5 mmHg) |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 113622 |
| Solubility Information | Very soluble in water. |
| Formula Weight | 142.18 |
| Percent Purity | 98% |
| Physical Form | Crystals or Powder or Crystalline Powder or Flakes |
| Chemical Name or Material | 3-Thiopheneacetic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H StatementH314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 230-166-1 |
| RTECSNumber | XM7570000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only