missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneacetic acid, 98%

Catalog No. AAA1070006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1070006 5 g
AAA1070014 25 g
AAA1070022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1070006 Supplier Thermo Scientific Chemicals Supplier No. A1070006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6964-21-2 | C6H6O2S | 142.172 g/mol

3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 6964-21-2
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD00005473
InChI Key RCNOGGGBSSVMAS-UHFFFAOYSA-N
Synonym 3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid
PubChem CID 23404
IUPAC Name 2-thiophen-3-ylacetic acid
SMILES C1=CSC=C1CC(=O)O

Specifications

Melting Point 77°C to 81°C
Boiling Point 129°C (2.5 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 113622
Solubility Information Very soluble in water.
Formula Weight 142.18
Percent Purity 98%
Chemical Name or Material 3-Thiopheneacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 230-166-1
RTECSNumber XM7570000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only