missing translation for 'onlineSavingsMsg'
Learn More

3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific™

Chemical Identifiers

CAS 775351-30-9
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
MDL Number MFCD11044436
InChI Key XMJKBDSKRITXBW-UHFFFAOYSA-N
Synonym 3-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, boronic acid,b-3-2h-tetrazol-5-yl phenyl, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, acmc-209pal
PubChem CID 46737996
IUPAC Name [3-(2H-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC=C1)C1=NNN=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445560010
SDS View Documents
Thermo Scientific Chemicals
445560010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

775351-30-9
189.97
XMJKBDSKRITXBW-UHFFFAOYSA-N
46737996
OB(O)C1=CC(=CC=C1)C1=NNN=N1
C7H7BN4O2
MFCD11044436
3-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, boronic acid,b-3-2h-tetrazol-5-yl phenyl, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, acmc-209pal
[3-(2H-tetrazol-5-yl)phenyl]boronic acid

Specifications

775351-30-9
Glass Bottle
MFCD11044436
3-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, boronic acid,b-3-2h-tetrazol-5-yl phenyl, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, acmc-209pal
OB(O)C1=CC(=CC=C1)C1=NNN=N1
189.97
189.97
3-(Tetrazol-5-yl)phenylboronic acid
95% min. (HPLC)
C7H7BN4O2
1g
XMJKBDSKRITXBW-UHFFFAOYSA-N
[3-(2H-tetrazol-5-yl)phenyl]boronic acid
46737996
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only