CAS RN 775351-30-9
IUPAC Name:
[3-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
Synonyms:
3-(2H-1,2,3,4-tetrazol-5-yl)phenylboronic acid
Molecular Weight (g/mol):
189.97
Molecular Formula:
C7H7BN4O2
InChi Key:
XMJKBDSKRITXBW-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=CC=C1)C1=NNN=N1
1
–
1
of
1
results
1
Promotions Available
3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals
3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals
CAS: 775351-30-9 Molecular Formula: C7H7BN4O2 Molecular Weight (g/mol): 189.97 MDL Number: MFCD11044436 InChI Key: XMJKBDSKRITXBW-UHFFFAOYSA-N Synonym: 3-tetrazol-5-yl phenylboronic acid,3-1h-tetrazol-5-yl phenylboronic acid,3-2h-tetrazol-5-yl-phenyl-boronic acid,3-2h-tetrazol-5-yl phenyl boronic acid,boronic acid,b-3-2h-tetrazol-5-yl phenyl,3-2h-tetrazol-5-yl phenyl-boronic acid,3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid,3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid,acmc-209pal PubChem CID: 46737996 IUPAC Name: [3-(2H-tetrazol-5-yl)phenyl]boronic acid SMILES: OB(O)C1=CC(=CC=C1)C1=NNN=N1