missing translation for 'onlineSavingsMsg'
Learn More

3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals

Catalog No. AC445560010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445560010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445560010 Supplier Thermo Scientific Chemicals Supplier No. 445560010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 775351-30-9 | C7H7BN4O2 | 189.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 775351-30-9
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
MDL Number MFCD11044436
InChI Key XMJKBDSKRITXBW-UHFFFAOYSA-N
Synonym 3-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, boronic acid,b-3-2h-tetrazol-5-yl phenyl, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, acmc-209pal
PubChem CID 46737996
IUPAC Name [3-(2H-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC=C1)C1=NNN=N1

Specifications

Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.97
Percent Purity 96%
Chemical Name or Material 3-(Tetrazol-5-yl)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only