missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butyltoluene 98.0+%, TCI America™

Catalog No. p-7118771
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09671G 1 g
B09675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09671G Supplier TCI America Supplier No. B09671G

Chemical Identifiers

CAS 1075-38-3
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00059210
InChI Key JTIAYWZZZOZUTK-UHFFFAOYSA-N
Synonym 3-tert-butyltoluene, toluene, m-tert-butyl, benzene, 1-1,1-dimethylethyl-3-methyl, 1-tert-butyl-3-methylbenzene, m-tert-butyltoluene, 1-methyl-3-tert-butylbenzene, butyltoluol, acmc-1bo1t, 1-methyl-3-tert-butyl-benzene, jtiaywzzzozutk-uhfffaoysa
PubChem CID 33711
IUPAC Name 1-tert-butyl-3-methylbenzene
SMILES CC1=CC(=CC=C1)C(C)(C)C

Specifications

Melting Point -41°C
Color Yellow
Boiling Point 189°C
Quantity 1 g
UN Number 2667
Formula Weight 148.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-tert-Butyltoluene
EINECSNumber (3)-0015
RTECSNumber XS8225000
TSCA No