missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylsalicylaldehyde 95.0+%, TCI America™

Catalog No. B40161G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40161G 1 g
B40165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B40161G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B40161G

Chemical Identifiers

CAS 24623-65-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00778877
InChI Key ROILLNJICXGZQQ-UHFFFAOYSA-N
Synonym 3-tert-butyl-2-hydroxybenzaldehyde, 3-tert-butylsalicylaldehyde, 3-t-butylsalicylaldehyde, benzaldehyde, 3-1,1-dimethylethyl-2-hydroxy, 3-tert-butyl-2-hydroxy-benzaldehyde, 3-tert-butyl salicylaldehyde, acmc-1ceew, 3-tertbutylsalicylaldehyde, 2-hydroxy-3-tert.butylbenzaldehyde, 3-tert-butyl-2-oxidanyl-benzaldehyde
PubChem CID 3678675
IUPAC Name 3-tert-butyl-2-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC=CC(C=O)=C1O

Specifications

Color Yellow
Boiling Point 79°C
Quantity 1 g
Formula Weight 178.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-tert-Butylsalicylaldehyde