CAS RN 24623-65-2
IUPAC Name:
3-tert-butyl-2-hydroxybenzaldehyde
Synonyms:
3-tert-butyl-2-hydroxybenzaldehyde
Molecular Weight (g/mol):
178.23
Molecular Formula:
C11H14O2
InChi Key:
ROILLNJICXGZQQ-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=CC(C=O)=C1O
1
–
1
of
1
results
1
3-tert-Butylsalicylaldehyde 95.0+%, TCI America™
CAS: 24623-65-2 Molecular Formula: C11H14O2 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00778877 InChI Key: ROILLNJICXGZQQ-UHFFFAOYSA-N Synonym: 3-tert-butyl-2-hydroxybenzaldehyde,3-tert-butylsalicylaldehyde,3-t-butylsalicylaldehyde,benzaldehyde, 3-1,1-dimethylethyl-2-hydroxy,3-tert-butyl-2-hydroxy-benzaldehyde,3-tert-butyl salicylaldehyde,acmc-1ceew,3-tertbutylsalicylaldehyde,2-hydroxy-3-tert.butylbenzaldehyde,3-tert-butyl-2-oxidanyl-benzaldehyde PubChem CID: 3678675 IUPAC Name: 3-tert-butyl-2-hydroxybenzaldehyde SMILES: CC(C)(C)C1=CC=CC(C=O)=C1O