missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsilyloxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B37261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37261G Supplier TCI America Supplier No. B37261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-(tert-Butyldimethylsilyloxy)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 261621-12-9
Melting Point 91°C
Color White-Yellow
Molecular Formula C12H21BO3Si
Quantity 1 g
MDL Number MFCD03701505
Synonym 3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid, 3-tert-butyldimethylsiloxy benzeneboronic acid, 3-tert-butyldimethylsilyl oxy phenylboronic acid, 3-tert-butyl dimethyl silyl oxyphenyl boronic acid, m-t-butyldimethylsilyloxy phenylboronic acid, 3-t-butyl dimethylsiloxy phenyl boronic acid
InChI Key RDQWADDNQONTLB-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C)(C)OC1=CC=CC(=C1)B(O)O
IUPAC Name {3-[(tert-butyldimethylsilyl)oxy]phenyl}boronic acid
Molecular Weight (g/mol) 252.19
PubChem CID 3463029
Formula Weight 252.19
Physical Form Crystalline Powder
Show More Show Less