missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsiloxy)benzeneboronic acid, tech. 90%

Catalog No. p-7030394
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2837803 1 g
AAH2837806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2837803 Supplier Thermo Scientific Chemicals Supplier No. H2837803
Only null left

CAS: 261621-12-9 | C12H21BO3Si | 252.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 261621-12-9
Molecular Formula C12H21BO3Si
Molecular Weight (g/mol) 252.19
MDL Number MFCD03701505
InChI Key RDQWADDNQONTLB-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid, 3-tert-butyldimethylsiloxy benzeneboronic acid, 3-tert-butyldimethylsilyl oxy phenylboronic acid, 3-tert-butyl dimethyl silyl oxyphenyl boronic acid, m-t-butyldimethylsilyloxy phenylboronic acid, 3-t-butyl dimethylsiloxy phenyl boronic acid
PubChem CID 3463029
IUPAC Name [3-[tert-butyl(dimethyl)silyl]oxyphenyl]boronic acid
SMILES CC(C)(C)[Si](C)(C)OC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 86°C to 91°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 252.19
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-(tert-Butyldimethylsiloxy)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only