missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylaniline 98.0+%, TCI America™

Catalog No. B40721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40721G 1 g
B40725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40721G Supplier TCI America Supplier No. B40721G

Chemical Identifiers

CAS 5369-19-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00125078
InChI Key DPKTVUKEPNBABS-UHFFFAOYSA-N
Synonym 3-tert-butyl aniline, 3-tert-butylbenzenamine, benzenamine, 3-1,1-dimethylethyl, 3-t-butyl aniline, 3-tert-butyl-phenylamine, 3-tert-butyl phenylamine, 3-tert-butylphenyl amine, m-t-butylaniline, 3-t-butylaniline, m-tert-butylaniline
PubChem CID 79334
IUPAC Name 3-tert-butylaniline
SMILES CC(C)(C)C1=CC=CC(N)=C1

Specifications

Color Yellow
Boiling Point 119°C
Quantity 1 g
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-tert-Butylaniline
EINECSNumber (3)-0206
TSCA No