missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butoxycarbonylamino)propyl Bromide 98.0+%, TCI America™

Catalog No. B22341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22341G 1 g
B22345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22341G Supplier TCI America Supplier No. B22341G

Chemical Identifiers

CAS 83948-53-2
Molecular Formula C8H16BrNO2
Molecular Weight (g/mol) 238.13
MDL Number MFCD02683429
InChI Key IOKGWQZQCNXXLD-UHFFFAOYSA-N
Synonym 3-boc-amino propyl bromide, tert-butyl 3-bromopropylcarbamate, tert-butyl n-3-bromopropyl carbamate, 3-boc-amino propylbromide, tert-butyl 3-bromopropyl carbamate, 3-bromo-propyl-carbamic acid tert-butyl ester, n-3-bromopropyl carbamic acid tert-butyl ester, 3-tert-butoxycarbonylamino propyl bromide, n-boc-3-bromopropylamine, 1-n-boc-3-bromopropylamine
PubChem CID 4460490
IUPAC Name tert-butyl N-(3-bromopropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCBr

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 1 g
Formula Weight 238.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(tert-Butoxycarbonylamino)propyl Bromide
TSCA No
Recommended Storage Freezer