accessibility menu, dialog, popup

UserName

H3CCH3H3COOHNBr

CAS RN 83948-53-2

IUPAC Name: tert-butyl N-(3-bromopropyl)carbamate
Synonyms: tert-butyl N-(3-bromopropyl)carbamate
Molecular Weight (g/mol): 238.13
Molecular Formula: C8H16BrNO2
InChi Key: IOKGWQZQCNXXLD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCBr
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)