missing translation for 'onlineSavingsMsg'
Learn More

3-[(tert-Butoxycarbonyl)amino]phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B42791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42791G Supplier TCI America Supplier No. B42791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-[(tert-Butoxycarbonyl)amino]phenylboronic Acid (contains varying amounts of Anhydride)
CAS 380430-68-2
Melting Point 168°C
Molecular Formula C11H16BNO4
Quantity 1 g
MDL Number MFCD03411945
Synonym 3-boc-aminophenyl boronic acid, 3-n-boc-amino phenylboronic acid, 3-tert-butoxycarbonylamino phenylboronic acid, n-boc-3-aminophenylboronic acid, 3-tert-butoxycarbonyl amino phenylboronic acid, 3-boronoaniline, n-boc protected, 3-tert-butoxycarbonyl amino benzeneboronic acid, 3-boc-amino benzeneboronic acid, 3-tert-butoxycarbonylamino phenyl-boronic acid, 3-tert-butoxycarbonyl amino phenyl boronic acid
InChI Key CWLNHPXWZRALFS-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)NC(=O)OC(C)(C)C)(O)O
IUPAC Name [3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
Molecular Weight (g/mol) 237.062
PubChem CID 2773228
Formula Weight 237.06
Physical Form Crystalline Powder
Show More Show Less