missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfobenzoic Acid Monosodium Salt 95.0+%, TCI America™

Catalog No. p-7135826
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S012625G 25 g
S0126500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S012625G Supplier TCI America Supplier No. S012625G
Only null left

Chemical Identifiers

CAS 17625-03-5
Molecular Formula C7H5NaO5S
Molecular Weight (g/mol) 224.162
MDL Number MFCD00066378
InChI Key KQHKITXZJDOIOD-UHFFFAOYSA-M
PubChem CID 23668512
IUPAC Name sodium;3-sulfobenzoate
SMILES C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Synonym sodium 3-sulfobenzoate, 3-sulfobenzoic acid monosodium salt, benzoic acid, 3-sulfo-, monosodium salt, benzoic acid, 3-sulfo-, sodium salt 1:1, sodium hydrogen m-sulphonatobenzoate, sodium3-sulfobenzoate, sodium m-sulfobenzoate, acmc-209sh5, 3-sodiosulfo benzoic acid, ksc491e3l
Formula Weight 224.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Sulfobenzoic Acid Monosodium Salt
EINECSNumber (3)-2132
TSCA Yes