missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfo-N-succinimidyl 4-(N-Maleimidomethyl)cyclohexane-1-carboxylate Sodium Salt, TCI America™

Catalog No. S088320MG
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S088320MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S088320MG Supplier TCI America Supplier No. S088320MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92921-24-9
Molecular Formula C16H17N2NaO9S
Molecular Weight (g/mol) 436.367
MDL Number MFCD00054978
InChI Key VUFNRPJNRFOTGK-UHFFFAOYSA-M
Synonym sulfo-smcc, sodium 1-4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarbonyl oxy-2,5-dioxopyrrolidine-3-sulfonate, sulfo-smcc sodium, 4-maleimidomethyl cyclohexane-1-carboxylic acid 3-sulfo-n-hydroxysuccinimide ester sodium salt, 3-sulfo-n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate sodium salt, sulfo-trans-smcc, sulfo-smcc crosslinker, sulfo-n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate sodium salt, sulfo-smcc sodium salt, bicl207
PubChem CID 4618794
IUPAC Name sodium;1-[4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexanecarbonyl]oxy-2,5-dioxopyrrolidine-3-sulfonate
SMILES C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CC(C3=O)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 20 mg
Formula Weight 436.37
Physical Form Crystalline Powder
Chemical Name or Material 3-Sulfo-N-succinimidyl 4-(N-Maleimidomethyl)cyclohexane-1-carboxylate Sodium Salt
TSCA No
Recommended Storage Freezer