accessibility menu, dialog, popup

UserName

CAS RN 92921-24-9

IUPAC Name: sodium 1-{4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexanecarbonyloxy}-2,5-dioxopyrrolidine-3-sulfonate
Synonyms: sodium 1-{4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexanecarbonyloxy}-2,5-dioxopyrrolidine-3-sulfonate
Molecular Weight (g/mol): 436.37
Molecular Formula: C16H17N2NaO9S
InChi Key: VUFNRPJNRFOTGK-UHFFFAOYNA-M
SMILES: [Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)C2CCC(CN3C(=O)C=CC3=O)CC2)C1=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)