missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 3-Quinolinol | |
| 203°C | |
| MFCD00169018 | |
| 3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r | |
| C1=CC=C2C(=C1)C=C(C=N2)O | |
| 145.161 | |
| 145.16 | |
| Crystalline Powder |
| 580-18-7 | |
| C9H7NO | |
| 5 g | |
| IQQDNMHUOLMLNJ-UHFFFAOYSA-N | |
| quinolin-3-ol | |
| 11376 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 580-18-7 | |
| 145.161 | |
| IQQDNMHUOLMLNJ-UHFFFAOYSA-N | |
| 11376 | |
| C1=CC=C2C(=C1)C=C(C=N2)O |
| C9H7NO | |
| MFCD00169018 | |
| 3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r | |
| quinolin-3-ol |
Safety and Handling
RTECSNumber : VC4050000
TSCA : No