missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Quinolinol 98.0+%, TCI America™
$113.71 - $354.17
Chemical Identifiers
| CAS | 580-18-7 |
|---|---|
| Molecular Formula | C9H7NO |
| Molecular Weight (g/mol) | 145.161 |
| MDL Number | MFCD00169018 |
| InChI Key | IQQDNMHUOLMLNJ-UHFFFAOYSA-N |
| Synonym | 3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r |
| PubChem CID | 11376 |
| IUPAC Name | quinolin-3-ol |
| SMILES | C1=CC=C2C(=C1)C=C(C=N2)O |
Chemical Identifiers
| 580-18-7 | |
| 145.161 | |
| IQQDNMHUOLMLNJ-UHFFFAOYSA-N | |
| 11376 | |
| C1=CC=C2C(=C1)C=C(C=N2)O |
| C9H7NO | |
| MFCD00169018 | |
| 3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r | |
| quinolin-3-ol |
Specifications
| 580-18-7 | |
| C9H7NO | |
| 1 g | |
| IQQDNMHUOLMLNJ-UHFFFAOYSA-N | |
| quinolin-3-ol | |
| 11376 | |
| ≥98.0% (GC,T) | |
| 3-Quinolinol |
| 203°C | |
| MFCD00169018 | |
| 3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r | |
| C1=CC=C2C(=C1)C=C(C=N2)O | |
| 145.161 | |
| 145.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : VC4050000
TSCA : No