missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Quinolinecarboxaldehyde 96.0+%, TCI America™
$159.79 - $540.97
Chemical Identifiers
| CAS | 13669-42-6 |
|---|---|
| Molecular Formula | C10H7NO |
| Molecular Weight (g/mol) | 157.172 |
| MDL Number | MFCD00006768 |
| InChI Key | RYGIHSLRMNXWCN-UHFFFAOYSA-N |
| Synonym | 3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde |
| PubChem CID | 83641 |
| IUPAC Name | quinoline-3-carbaldehyde |
| SMILES | C1=CC=C2C(=C1)C=C(C=N2)C=O |
Chemical Identifiers
| 13669-42-6 | |
| 157.172 | |
| RYGIHSLRMNXWCN-UHFFFAOYSA-N | |
| 83641 | |
| C1=CC=C2C(=C1)C=C(C=N2)C=O |
| C10H7NO | |
| MFCD00006768 | |
| 3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde | |
| quinoline-3-carbaldehyde |
Specifications
| 13669-42-6 | |
| Yellow | |
| C10H7NO | |
| 1 g | |
| RYGIHSLRMNXWCN-UHFFFAOYSA-N | |
| quinoline-3-carbaldehyde | |
| 83641 | |
| ≥96.0% (GC,T) | |
| 3-Quinolinecarboxaldehyde |
| 67°C | |
| 128°C | |
| MFCD00006768 | |
| 3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde | |
| C1=CC=C2C(=C1)C=C(C=N2)C=O | |
| 157.172 | |
| 157.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No