missing translation for 'onlineSavingsMsg'
Learn More

3-Quinolinecarboxaldehyde 96.0+%, TCI America™

Catalog No. Q00755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00755G 5 g
Q00751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00755G Supplier TCI America Supplier No. Q00755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13669-42-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00006768
InChI Key RYGIHSLRMNXWCN-UHFFFAOYSA-N
Synonym 3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
PubChem CID 83641
IUPAC Name quinoline-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(C=N2)C=O

Specifications

Melting Point 67°C
Color Yellow
Boiling Point 128°C
Quantity 5 g
Formula Weight 157.17
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Quinolinecarboxaldehyde