missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyrroline Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P22401G
Specifications
| 3-Pyrroline Hydrochloride | |
| 172°C | |
| C4H8ClN | |
| 1 g | |
| GBGHNSYFGCVGDX-UHFFFAOYSA-N | |
| 2,5-dihydro-1H-pyrrole;hydrochloride | |
| 11679809 | |
| ≥98.0% (T) |
| 63468-63-3 | |
| Yellow | |
| MFCD09991892 | |
| 2,5-Dihydro-1H-pyrrole Hydrochloride | |
| C1C=CCN1.Cl | |
| 105.565 | |
| 105.57 | |
| Crystalline Powder |
Chemical Identifiers
| 63468-63-3 | |
| 105.565 | |
| GBGHNSYFGCVGDX-UHFFFAOYSA-N | |
| 11679809 | |
| C1C=CCN1.Cl |
| C4H8ClN | |
| MFCD09991892 | |
| 2,5-Dihydro-1H-pyrrole Hydrochloride | |
| 2,5-dihydro-1H-pyrrole;hydrochloride |
Safety and Handling
TSCA : No