accessibility menu, dialog, popup

UserName

CAS RN 63468-63-3

IUPAC Name: 2,5-dihydro-1H-pyrrole hydrochloride
Synonyms: 3-pyrroline hydrochloride
Molecular Weight (g/mol): 105.57
Molecular Formula: C4H8ClN
InChi Key: GBGHNSYFGCVGDX-UHFFFAOYSA-N
SMILES: Cl.C1NCC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results