missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyridylacetic Acid Hydrochloride 99.0+%, TCI America™
$150.98 - $300.11
Chemical Identifiers
| CAS | 6419-36-9 |
|---|---|
| Molecular Formula | C7H8ClNO2 |
| Molecular Weight (g/mol) | 173.596 |
| MDL Number | MFCD00012819 |
| InChI Key | XVCCOEWNFXXUEV-UHFFFAOYSA-N |
| Synonym | 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride |
| PubChem CID | 2723724 |
| IUPAC Name | 2-pyridin-3-ylacetic acid;hydrochloride |
| SMILES | C1=CC(=CN=C1)CC(=O)O.Cl |
Chemical Identifiers
| 6419-36-9 | |
| 173.596 | |
| XVCCOEWNFXXUEV-UHFFFAOYSA-N | |
| 2723724 | |
| C1=CC(=CN=C1)CC(=O)O.Cl |
| C7H8ClNO2 | |
| MFCD00012819 | |
| 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride | |
| 2-pyridin-3-ylacetic acid;hydrochloride |
Specifications
| 6419-36-9 | |
| White-Yellow | |
| MFCD00012819 | |
| 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride | |
| C1=CC(=CN=C1)CC(=O)O.Cl | |
| 173.596 | |
| 173.60 | |
| Crystalline Powder |
| 160°C | |
| C7H8ClNO2 | |
| 10 g | |
| XVCCOEWNFXXUEV-UHFFFAOYSA-N | |
| 2-pyridin-3-ylacetic acid;hydrochloride | |
| 2723724 | |
| ≥99.0% (T) | |
| 3-Pyridylacetic Acid Hydrochloride |
Safety and Handling
EINECSNumber : (5)-3711
TSCA : No