accessibility menu, dialog, popup

UserName

OON

CAS RN 6419-36-9

IUPAC Name: 2-(pyridin-3-yl)acetate
Synonyms: 3-pyridylacetate
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H6NO2
InChi Key: WGNUNYPERJMVRM-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)