missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridylacetic Acid Hydrochloride 99.0+%, TCI America™

Catalog No. P063925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P063925G 25 g
P063910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P063925G Supplier TCI America Supplier No. P063925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6419-36-9
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012819
InChI Key XVCCOEWNFXXUEV-UHFFFAOYSA-N
Synonym 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride
PubChem CID 2723724
IUPAC Name 2-pyridin-3-ylacetic acid;hydrochloride
SMILES C1=CC(=CN=C1)CC(=O)O.Cl

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 25 g
Formula Weight 173.60
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Pyridylacetic Acid Hydrochloride
EINECSNumber (5)-3711
TSCA No