missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7135421
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21311G 1 g
P21315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21311G Supplier TCI America Supplier No. P21311G
Only null left

Chemical Identifiers

CAS 17452-27-6
Molecular Formula C6H4N2S
Molecular Weight (g/mol) 136.172
MDL Number MFCD00052351
InChI Key VMSZFBSYWXMXRF-UHFFFAOYSA-N
PubChem CID 2737242
IUPAC Name 3-isothiocyanatopyridine
SMILES C1=CC(=CN=C1)N=C=S

Specifications

Color Yellow
Boiling Point 65°C
Quantity 1 g
Synonym 3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
Formula Weight 136.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pyridyl Isothiocyanate
TSCA No
Recommended Storage Refrigerator