missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyridinemethanol 98.0+%, TCI America™
Supplier: TCI America P0555500ML
Specifications
| 3-Pyridinemethanol | |
| Yellow | |
| C6H7NO | |
| 500 mL | |
| MVQVNTPHUGQQHK-UHFFFAOYSA-N | |
| (pyridin-3-yl)methanol | |
| 7510 | |
| ≥98.0% (GC) |
| 100-55-0 | |
| 145°C | |
| MFCD00006407 | |
| 3-pyridinemethanol, nicotinyl alcohol, 3-hydroxymethyl pyridine, 3-pyridylmethanol, nicotinic alcohol, 3-pyridylcarbinol, roniacol, pyridylcarbinol, 3-pyridinylmethanol, pyridine-3-carbinol | |
| OCC1=CC=CN=C1 | |
| 109.13 | |
| 109.13 | |
| Liquid |
Chemical Identifiers
| 100-55-0 | |
| 109.13 | |
| MVQVNTPHUGQQHK-UHFFFAOYSA-N | |
| 7510 | |
| OCC1=CC=CN=C1 |
| C6H7NO | |
| MFCD00006407 | |
| 3-pyridinemethanol, nicotinyl alcohol, 3-hydroxymethyl pyridine, 3-pyridylmethanol, nicotinic alcohol, 3-pyridylcarbinol, roniacol, pyridylcarbinol, 3-pyridinylmethanol, pyridine-3-carbinol | |
| (pyridin-3-yl)methanol |
Safety and Handling
EINECSNumber : (5)-0727
RTECSNumber : UT4690000
TSCA : Yes