missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinemethanol 98.0+%, TCI America™

Catalog No. p-7134673
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P055525ML 25 mL
P0555500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P055525ML Supplier TCI America Supplier No. P055525ML
Only null left

Chemical Identifiers

CAS 100-55-0
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006407
InChI Key MVQVNTPHUGQQHK-UHFFFAOYSA-N
Synonym 3-pyridinemethanol, nicotinyl alcohol, 3-hydroxymethyl pyridine, 3-pyridylmethanol, nicotinic alcohol, 3-pyridylcarbinol, roniacol, pyridylcarbinol, 3-pyridinylmethanol, pyridine-3-carbinol
PubChem CID 7510
IUPAC Name (pyridin-3-yl)methanol
SMILES OCC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
Formula Weight 109.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pyridinemethanol
EINECSNumber (5)-0727
RTECSNumber UT4690000
TSCA Yes